Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLN13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548962

Catalog No. PA548962

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

SLN13
Polyclonal
PBS with 0.09% sodium azide
Q68D06
Schlafen family member 13, SLFN13
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
146857
Western Blot
Unconjugated
SLFN13
Liquid
SLFN13
KLH conjugated synthetic peptide between 632-663 amino acids from the C-terminal region of human SLN13.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit