Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slo3 Mouse anti-Human, Mouse, Rat, Clone: S2-16, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527598

Catalog No. PIMA527598


Add to cart

Description

Description

The Slo3 channel is a novel potassium channel abundantly expressed in mammalian speramtocytes- tests have shown that it is expressed in both mouse and human testis. It represents a new and unique type of potassium channel that is regulated by both intracellular pH and membrane voltage. Because of its sensitivity to both pH and voltage, Slo3 may play a role in alkalization-mediated K(+) fluxes associated with sperm capacitation.
Specifications

Specifications

Slo3
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
A8MYU2, O54982
Kcnma3, mSlo3, Slo3, KCa5, KCNMA3, potassium channel subfamily U member 1, Slowpoke homolog 3, KCa5.1, KCNMC1Calcium-activated potassium channel, subfamily M subunit alpha-3, Calcium-activated potassium channel subunit alpha-3, potassium channel, subfamily U, member 1
Mouse
IgG2b
100 μg
-20°C
Primary
157855, 16532, 680912
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
S2-16
Unconjugated
KCNU1
Liquid
Kcnu1
Fusion protein amino acids 1052-1121 of mouse Slo3
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit