Learn More
Eisco™ Sloping Splash Heads - Pear Shaped Products for Science Education
Typically used in distillation assemblies to prevent the carry-over raw liquid into the vapor stream. A variety of flask and condenser cone sizes are available.
Manufacturer:  Eisco™ CH0856B
 View more versions of this product
Catalog No. S29490
Add to cart
Description
Description
  • Sloping splash head adapter
  • Pear shaped trap
  • For used to prevent carry over of raw liquids into the vapor steam
  • Made from high quality borosilicate 3.3 glass
  • Autoclavable
  • Specifications
    Specifications
    Distillation
    Borosilicate Glass 3.3
    To fit flask- 24/29, To fit condenser- 19/26
    Sloping
    1
    SDS
    Documents
    Provide Content Correction
    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Cancel Submit
    1 year limited manufacturer warranty