Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Slotted Brass Weight Set with Hanger Products for Science Education

Removable, brass slotted weight set with hanger is ideal for a variety of physics demonstrations and experiments.

$14.75 - $81.00

Specifications

Description Slotted
For Use With (Application) Physics Demonstrations and Experiments
Material Brass
Product Type Weight Set
View More Specs

Products 4
Catalog Number Mfr. No. Mass Price Quantity & Availability  
Catalog Number Mfr. No. Mass Price Quantity & Availability  
S15111 Products for Science Education
View Documents
Eisco™
PH0258A
10 g
Each for $14.75
Add to cart
 
S15112 Products for Science Education
View Documents
Eisco™
PH0258B
25 g
Each for $25.75
Add to cart
 
S15113 Products for Science Education
View Documents
Eisco™
PH0258C
50 g
Each for $23.50
Add to cart
 
S15114 Products for Science Education
View Documents
Eisco™
PH0258D
100 g
Each for $81.00
Add to cart
 
Description

Description

  • Sets of removable brass masses with slots
  • Brass hanger with hook
  • Fine, smooth finish
Specifications

Specifications

Slotted
Physics Demonstrations and Experiments
Brass
Weight Set
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit