Learn More
Slow Skeletal Muscle Myosin Heavy Chain Mouse anti-Human, Mouse, Rat, Chicken, Rabbit, Janelia Fluor 646, Clone: 9812, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250299JF646
Catalog No. NBP50299646
Add to cart
Description
Description
Slow Skeletal Muscle Myosin Heavy Chain Monoclonal antibody specifically detects Slow Skeletal Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Immunohistochemistry.
Specifications
Specifications
Slow Skeletal Muscle Myosin Heavy Chain
9812
Immunohistochemistry
Purified
Purified human skeletal muscle myosin
Protein G purified
Store at 4°C in the dark.
Monoclonal
Chicken, Human, Mouse, Rabbit, Rat
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1
0.1mL
Primary
Specific for slow skeletal muscle myosin heavy chain (SM1).