Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLTM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116846
Description
Positive Samples: Mouse brain Immunogen sequence: RPERSGREVS GHSVRGAPPG NRSSASGYGS REGDRGVITD RGGGSQHYPE ERHVVERHGR DTSGPRKEWH GPPSQGPSYH DTRRMGDGRA GAGMITQHSS N.
SLTM gene ontology annotations related to this gene include RNA binding.
Specifications
SLTM | |
Polyclonal | |
Unconjugated | |
SLTM | |
5730455C01Rik; 5730555F13Rik; 9130215G10Rik; FLJ13213; Met; modulator of estrogen induced transcription; modulator of estrogen-induced transcription; RGD1307526; SAFB like transcription modulator; SAFB-like; SAFB-like transcription modulator; SAFB-like, transcription modulator; SAF-like transcription modulator; SLTM | |
Rabbit | |
Affinity chromatography | |
RUO | |
66660 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.26 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8CH25 | |
SLTM | |
A synthetic peptide corresponding to a sequence within amino acids 900-1000 of human SLTM (NP_0790312). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction