Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLU7 Mouse anti-Human, DyLight 680, Clone: 262, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243793FR

Catalog No. NBP243793FR


Add to cart

Description

Description

SLU7 Monoclonal antibody specifically detects SLU7 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SLU7
262
Western Blot
Purified
SLU7
Recombinant protein encompassing a sequence within the center region of human SLU7. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
hSlu7, MGC9280, pre-mRNA-splicing factor SLU7,9G8, SLU7 splicing factor homolog (S. cerevisiae), splicing factor, step II splicing factor SLU7, zinc knuckle motif containing
Mouse
IgG2a
0.1 ml
Primary
10569
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit