Tocris™ (±)-SLV 319

Potent and selective CB1 receptor antagonist

Manufacturer: Tocris™ 4605/50

 View more versions of this product

Catalog No. 46-055-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors (±)-SLV 319
    Purity >98
    Molecular Weight 487.4
    Quantity 50mg
    Product Type (±)-SLV 319

    Potent and selective CB1 receptor antagonist (Ki = 7.8nM). Exhibits 1000-fold selectivity for CB1 over CB2 receptors. Inhibits CP 55,940-induced hypotension and WIN 55,212-2-induced hypothermia in vivo. Orally active.

    • Chemical Name: 3-(4-Chlorophenyl)-N-[(4-Chlorophenyl)sulfonyl]-4,5-dihydro-N'-methyl-4-phenyl-1H-pyrazole-1-carboximidamide
    • Formula: C23H20Cl2N4O2S
    • CAS Number: 362519-49-1
    • Solubility: Soluble to 100mM in DMSO and to 10mM in ethanol with gentle warming
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit