Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLV 320, Tocris Bioscience™

Catalog Number 334450
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
251945-92-3
C18H20N4O
308.385
RBZNJGHIKXAKQE-UHFFFAOYSA-N
derenofylline, unii-o1q96uz63w, trans-4-2-phenyl-7h-pyrrolo 2,3-d pyrimidin-4-yl amino cyclohexanol, derenofylline inn, d0f5un, 4-4-phenyl-3,5,7-triazabicyclo 4.3.0 nona-2,4,8,10-tetraen-2-yl amino cyclohexan-1-ol, slv hplc, 4-2-phenyl-7h-pyrrolo 2,3-d pyrimidin-4-ylamino cyclohexanol, 4-2-phenyl-7h-pyrrolo 2,3-d pyrimidin-4-yl amino cyclohexan-1-ol, cyclohexanol, 4-2-phenyl-1h-pyrrolo 2,3-d pyrimidin-4-yl amino-, trans
9953065
4-[(2-phenyl-7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]cyclohexan-1-ol
C1CC(CCC1NC2=NC(=NC3=C2C=CN3)C4=CC=CC=C4)O
This item is not returnable. View return policy
251945-92-3
C18H20N4O
308.385
RBZNJGHIKXAKQE-UHFFFAOYSA-N
derenofylline, unii-o1q96uz63w, trans-4-2-phenyl-7h-pyrrolo 2,3-d pyrimidin-4-yl amino cyclohexanol, derenofylline inn, d0f5un, 4-4-phenyl-3,5,7-triazabicyclo 4.3.0 nona-2,4,8,10-tetraen-2-yl amino cyclohexan-1-ol, slv hplc, 4-2-phenyl-7h-pyrrolo 2,3-d pyrimidin-4-ylamino cyclohexanol, 4-2-phenyl-7h-pyrrolo 2,3-d pyrimidin-4-yl amino cyclohexan-1-ol, cyclohexanol, 4-2-phenyl-1h-pyrrolo 2,3-d pyrimidin-4-yl amino-, trans
9953065
4-[(2-phenyl-7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]cyclohexan-1-ol
C1CC(CCC1NC2=NC(=NC3=C2C=CN3)C4=CC=CC=C4)O

Potent and selective A1 antagonist

Potent and selective adenosine A1 receptor antagonist (Ki values are 1, 200, 398 and 3981 nM at human A1, A3, A2A and A2B receptors respectively). Suppresses cardiac fibrosis and attenuates albuminuria, without effect on blood pressure in animal models of chronic renal failure. Orally active.
Quantity 50 mg
Formula Weight 308.38
Percent Purity >99%
Chemical Name or Material SLV 320
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit