Learn More
SLV 320, Tocris
Potent and selective A1 antagonist
Manufacturer:  R&D Systems 3344/10
Quantity 10mg
 View more versions of this product
Catalog No. 33-441-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
308.38
SLV 320
C18H20N4O
trans-4-[(2-Phenyl-7H-pyrrolo[2, 3-d]pyrimidin-4-yl)amino]cyclohexanol
Specifications
10mg
>99%
308.38
SLV 320
251945-92-3
C18H20N4O
trans-4-[(2-Phenyl-7H-pyrrolo[2, 3-d]pyrimidin-4-yl)amino]cyclohexanol
Safety and Handling
Recommended Storage : Store at Room Temperature