Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad4 Mouse anti-Human, Clone: SPM448, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30726310UG

 View more versions of this product

Catalog No. NB121999


Add to cart

Description

Description

Smad4 Monoclonal specifically detects Smad4 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Smad4
SPM448
Unconjugated
10 mM PBS with 0.05% BSA
DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4
Recombinant full-length human Smad4 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG2a Kappa
100 μg
Signal Transduction, Stem Cell Signaling Pathway
Primary
4089
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit