Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad4 Mouse anti-Human, Clone: SPM448, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Smad4
Clone SPM448
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122000
View Documents
Novus Biologicals
NBP30726320UG
20 μg
Each for $138.37
Add to cart
 
NB121999
View Documents
Novus Biologicals
NBP30726310UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Smad4 Monoclonal antibody specifically detects Smad4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Smad4
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4
Recombinant full-length human Smad4 protein
Monoclonal
Protein A or G purified
SPM448
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Signal Transduction, Stem Cell Signaling Pathway
10 mM PBS with 0.05% BSA
4089
IgG2a Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit