Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Smad4
Applications Immunofluorescence, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093630
View Documents
Novus Biologicals
NBP303495100UL
100 μg Each for $320.71
Add to cart
 
NB093631
View Documents
Novus Biologicals
NBP30349520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Smad4 Polyclonal antibody specifically detects Smad4 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence, KnockDown
Specifications

Specifications

Smad4
Unconjugated
Rabbit
Signal Transduction, Stem Cell Signaling Pathway
PBS with 50% glycerol, pH7.3.
4089
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, KnockDown, Western Blot
Purified
RUO
Human, Mouse
DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4
Recombinant fusion protein containing a sequence corresponding to amino acids 160-450 of human Smad4 (NP_005350.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit