Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Small Geiger Tube
SDPSCI_ED

End window is recessed about 2cm to prevent breakage.

Supplier:  Science First EN01

Catalog No. S440973

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.


Description

Description

  • Small 0.8 in. (1.5cm) Geiger tube
  • Stainless-steel tube
  • Tube is halogen quenched
  • Mica end window is 2.3mg/cm2 thick
  • Operating plateau: 450VDC

Specifications

Specifications

Geiger Tube
Stainless Steel
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit