Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Harvard Apparatus Small Nylon Clamps
Made of strong, lightweight nylon reinforced with glass fiber.
$111.00
Catalog Number | Mfr. No. | Includes | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Color | Price | Quantity & Availability | ||||
05-886-3
|
Harvard Apparatus
53 2102R |
Extension stem | Red |
Each for $111.00
|
|
Description
Choose red or white; integral color won't peel or flake.
- One moving jaw
- Open to 5/8 in. (16mm); close completely
- Resist most chemicals, particularly salt solutions and solvents
- Heat resistant to 221°C (430°F)
- Extension stem is 1/4 in. diameter (6.4mm) and 5 in. long (125mm)
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title