Learn More
Harvard Apparatus™ Small Nylon Vice Clamps
Securely holds thin sections such as PCBs, slides, rulers, filters, targets, boards, etc.
Manufacturer:  Harvard Apparatus™ 53 2122R
Catalog No. 05-886-13
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Parallel, straight jaws 1 in. wide (25mm); open to 1/3 in. (8mm). Close completely. Made of strong, lightweight nylon reinforced with glass fiber. Resist most chemicals, particularly salt solutions and solvents. Heat resistant to 221°C (430°F). For holding PCB's, slides, rulers, filters, targets, boards, etc. Mount on any 1/2 in. diameter (13mm) rod. Choose red or white; integral color won't peel or flake.
Specifications
Specifications
Vice/Board Clamp
0 to 0.33 in.
Small
430°F
BTX Harvard Apparatus
Nylon
0 to 0.83 cm
221°C
Red