Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotano Small Primate Elbow Length Catch Glove™

Premium suede leather gloves combine safety and dexterity to provide tactility, dexterity, and puncture resistance for safer handling/restraint techniques. Designed for trained professionals working with small primates in Lab Animal Research Facilities.

$3042.85 - $7608.57

Specifications

Color Tan
For Use With (Application) Protection Against Bites, Scratches, Punctures, and Lacerations When Handling Primates and Other Animals
Glove Cuff Style Gauntlet
Grade Premium
Material Suede Leather
View More Specs

Products 9
Catalog Number Mfr. No. Hand Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Hand Quantity Price Quantity & Availability  
17-116-025
View Documents
Rotano
NBSPOFALH20
Left 20 Gloves/Cs
Case for $3,042.85
Add to cart
 
17-116-022
View Documents
Rotano
NBSPOFALH30
Left 30 Gloves/Cs
Case for $4,562.85
Add to cart
 
17-116-019
View Documents
Rotano
NBSPOFALH50
Left 50 Gloves/Cs
Case for $7,608.57
Add to cart
 
17-116-024
View Documents
Rotano
NBSPOFARH20
Right 20 Gloves/Cs
Case for $3,042.85
Add to cart
 
17-116-021
View Documents
Rotano
NBSPOFARH30
Right 30 Gloves/Cs
Case for $4,562.85
Add to cart
 
17-116-018
View Documents
Rotano
NBSPOFARH50
Right 50 Gloves/Cs
Case for $7,608.57
Add to cart
 
17-116-026
View Documents
Rotano
NBSPOFAPR10
Specific 10 Pair/Cs
Case for $3,042.85
Add to cart
 
17-116-023
View Documents
Rotano
NBSPOFAPR15
Specific 15 Pair/Cs
Case for $4,562.85
Add to cart
 
17-116-020
View Documents
Rotano
NBSPOFAPR25
Specific 25 Pair/Cs
Case for $7,608.57
Add to cart
 
Description

Description

  • Gloves provide protection against bites, scratches, and lacerations when handling small primates
  • High dexterity, tactile sensitivity, and flexibility allow for proper and safe restraint techniques
  • Reinforced double layer of premium leather in the thumb and palm for additional protection
  • Fleeced poly cotton liner for added insulation, sweat absorption, and comfort
  • Leather thimbles on the inside of the forefinger and middle fingertips for added bite protection
  • Split seam design prevents any roll down, provides arm protection, and allows for greater movement when them arm is bent
  • High strength seam construction allows for added durability and strength
  • Non-exposed stitching protects against snagging
  • Tested to EN 388:2016 + A1:2018 6.5 Puncture Resistance, Specification Force (N): Performance Level 4
Specifications

Specifications

Tan
Gauntlet
Suede Leather
One Size
Keystone
Protection Against Bites, Scratches, Punctures, and Lacerations When Handling Primates and Other Animals
Premium
Animal Handling Gloves
NBSP-OFA
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit