Learn More
Erdco Engineering Corporation SMALL SCALE FLASHPOINT TESTER
SDP
Manufacturer:  Erdco Engineering Corporation RT00001
Encompass_Preferred
Test time is only one minute with a 2 ml sample for flash points below 100°C (212°F). You can run ten tests with the Rapid Tester® in the time that it takes to perform just one by other methods. The 2 ml/4 ml sample is a fraction of the large sample required by other flash point methods. This reduces sampling and disposal problems while improving safety. Digital circuitry permits simple adjustment and precise automatic regulation of test temperatures. Depress the Preset switch while turning the Temperature Control knob until the target temperature is displayed. Test cup temperature is automatically shown on the LCD display.
Catalog No. NC0783348