Learn More
Smallpox B5R Mouse anti-Virus, Alexa Fluor™ 532, Clone: 1J2F3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP224699AF532
Catalog No. NP224699A32
Add to cart
Description
Description
Smallpox B5R Monoclonal antibody specifically detects Smallpox B5R in Virus samples. It is validated for Western Blot.
Specifications
Specifications
Smallpox B5R
1J2F3
Western Blot
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Within the range of Amino acids 33-62 of the B5R protein were used as the immunogen.
0.1 mL
Primary
Virus