Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMAP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | SMAP1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155453
|
Novus Biologicals
NBP155453 |
100 μL |
Each of 1 for $436.00
|
|
Description
SMAP1 Polyclonal specifically detects SMAP1 in Human samples. It is validated for Western Blot.Specifications
SMAP1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ13159, FLJ42245, small ArfGAP 1, SMAP-1, stromal membrane-associated GTPase-activating protein 1, stromal membrane-associated protein 1 | |
SMAP1 | |
IgG | |
Affinity Purified | |
50 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q8IYB5 | |
60682 | |
Synthetic peptides corresponding to SMAP1(stromal membrane-associated GTPase-activating protein 1) The peptide sequence was selected from the middle region of SMAP1. Peptide sequence KEPEKPAKPLTAEKLQKKDQQLEPKKSTSPKKAAEPTVDLLGLDGPAVAP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title