Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAP2 Mouse anti-Human, Clone: OTI9B6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527417

Catalog No. PIMA527417


Add to cart

Description

Description

GTPase activating protein that acts on ARF1. Can also activate ARF6 (in vitro). May play a role in clathrin-dependent retrograde transport from early endosomes to the trans-Golgi network
Specifications

Specifications

SMAP2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WU79
SMAP2, SMAP1L
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64744
Immunohistochemistry (Paraffin), Western Blot
OTI9B6
Unconjugated
SMAP2
Liquid
SMAP2
Full length human recombit protein of SMAP2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit