Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen SMARCA1
Dilution Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry
Applications Western Blot, ChIP assay, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125843
View Documents
Novus Biologicals
NBP310471100UL
100 μg
Each for $320.71
Add to cart
 
NB125844
View Documents
Novus Biologicals
NBP31047125UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

SMARCA1 Polyclonal antibody specifically detects SMARCA1 in Human samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry
Specifications

Specifications

SMARCA1
Western Blot, ChIP assay, Immunohistochemistry
Purified
RUO
PBS buffer, 2% sucrose
6594
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry
Unconjugated
Rabbit
Human
ATP-dependent helicase SMARCA1, DKFZp686D1623, EC 3.6.1, EC 3.6.4.-, FLJ41547, global transcription activator homologous sequence, Nucleosome-remodeling factor subunit SNF2L, NURF140, probable global transcription activator SNF2L1, SNF2L, SNF2L1ISWI, SNF2LB, SNF2-like 1, SNF2LT, subfamily a, member 1, sucrose nonfermenting 2-like protein 1, SWI, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 1, SWI2
The immunogen is a synthetic peptide directed towards the N terminal region of human SMARCA1 (NP_003060). Peptide sequence EQDTAAVAATVAAADATATIVVIEDEQPGPSTSQEEGAAAAATEATAATE
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit