Learn More
SMARCA5/SNF2H Mouse anti-Human, Clone: C.15100040, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259190
Catalog No. NBP259190
Add to cart
Description
Description
SMARCA5/SNF2H Monoclonal antibody specifically detects SMARCA5/SNF2H in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunofluorescence.
Specifications
Specifications
SMARCA5/SNF2H
C.15100040
Western Blot 1:1000, Immunoprecipitation 1 ug, Immunofluorescence 1:100 - 1:500
Ascites
SMARCA5
The exact immunogen is propietary information.
Ascites
RUO
Primary
8467
Western Blot, Immunoprecipitation, Immunofluorescence
Unconjugated
Ascites Fluid with 0.05% Sodium Azide
EC 3.6.1, EC 3.6.4.-, hISWI, hSNF2HSWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5, ISWI, SNF2Hsucrose nonfermenting-like 5, subfamily a, member 5, Sucrose nonfermenting protein 2 homolog, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 5, WCRF135
Mouse
IgG2b
100 ul
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human