Learn More
SMARCAD1/ETL1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301593A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine
SMARCAD1 belongs to the SNF2/RAD54 helicase family. It contains 2 CUE domains, 1 helicase ATP-binding domain, and 1 helicase C-terminal domain. It is a probable ATP-dependent DNA helicase.Specifications
| SMARCAD1/ETL1 | |
| Polyclonal | |
| Unconjugated | |
| SMARCAD1 | |
| ADERM, ATP-dependent helicase 1, DKFZP762K2015, ETL1, HEL1, hHEL1, KIAA1122 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56916 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9H4L7 | |
| SMARCAD1 | |
| Between 976 and 1026 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.