Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCAL1 Polyclonal Antibody
SMARCAL1 Polyclonal Antibody
Supplier: Thermo Scientific PA528980
Description
SMARCAL1 Polyclonal Antibody for Western Blot, ICC/IF, IP

Specifications
SMARCAL1 | |
Polyclonal | |
Unconjugated | |
SMARCAL1 | |
6030401P21Rik; AI851716; ATP-driven annealing helicase; DNA-dependent ATPase A; HARP; HepA-related protein; HHARP; Mharp; SMARCAL1; SMARCA-like protein 1; Sucrose nonfermenting protein 2-like 1; Swi/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A-like protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
316477, 50485, 54380 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
1.07 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
B4F769, Q8BJL0, Q9NZC9 | |
SMARCAL1 | |
Recombinant fragment corresponding to a region within amino acids 120 and 457 of Human SmarcAL1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction