Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCB1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA125450
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA125450 100 μL
1 options

Catalog No. PIPA125450

Supplier: Thermo Scientific PA125450

Rabbit Polyclonal Antibody

PA1-25450 detects SMARCB1 from human samples. PA1-25450 is expected to cross react with mouse (100% conserved) and zebrafish (100% conserved) due to sequence homology. PA1-25450 has been successfully used in Western blot procedures. The PA1-25450 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues A(294) L K L C S E L G L G G E F(307) of human SMARCB1 with N-terminally added Cysteine. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is part of a complex that relieves repressive chromatin structures, allowing the transcriptional machinery to access its targets more effectively. The encoded nuclear protein may also bind to and enhance the DNA joining activity of HIV-1 integrase. This gene has been found to be a tumor suppressor, and mutations in it have been associated with maligt rhabdoid tumors. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCB1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene SMARCB1
Gene Accession No. Q12824, Q9Z0H3, Q5U379
Gene Alias BAF47, INI1, RDT, SNF5, SNF5L1, Sfh1p, Snr1, hSNFS
Gene Symbols SMARCB1, SMARCB1A
Host Species Rabbit
Immunogen Synthetic peptide, conjugated to KLH, corresponding to residues A(294) L K L C S E L G L G G E F(307) of human SMARCB1 with N-terminally added Cystein
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20587, 6598, 799716
Target Species Fish, Human, Mouse, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.