Learn More
Invitrogen™ SMARCB1 Recombinant Rabbit Monoclonal Antibody (8A7O2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA557894

Description
Immunogen sequence: LWRRLATVEE RKKIVASSHG KKTKPNTKDH GYTTLATSVT LLKASEVEEI LDGNDEKYKA VSISTEPPTY LREQKAKRNS QWVPTLPNSS HHLDAVPCST T.
The SWI-SNF complex is involved in the activation of transcription via the remodeling of nucleosome structure in an ATP-dependent manner. Brm (also designated SNF2alpha) and Brg-1 (also designated SNF2beta) are the ATPase subunits of the mammalian SWI-SNF complex. Brm, Brg-1, Ini1 (integrase interactor 1, also designated SNF5), BAF155 (also designated SRG3) and BAF170 are thought to comprise the functional core of the SWI-SNF complex. Addition of Ini1, BAF155 and BAF170 to Brg-1 appears to increase remodeling activity. Other complex subunits are thought to play regulatory roles. hSNF2L and hSNF2H both appear to be homologs of Drosophila ISWI, a Brm related ATPase that is present in chromatin remodeling complexes other than SWI/SNF, including the NURF (nucleosome remodeling factor).
Specifications
SMARCB1 | |
Recombinant Monoclonal | |
1.86 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q12824, Q9Z0H3 | |
SMARCB1 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human SMARCB1/SNF5 (NP_003064.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
8A7O2 | |
Unconjugated | |
SMARCB1 | |
AU020204; Baf47; BRG1-associated factor 47; BRG1-associated factor 47 (BAF47); CSS3; hSNF5; hSNFS; INI1; Integrase interactor 1 protein; malignant rhabdoid tumor suppressor; MRD15; mSNF5; PPP1R144; protein phosphatase 1, regulatory subunit 144; RDT; RTPS1; Sfh1p; SMARCB1; smarcb1a; SNF5; SNF5 homolog; SNF5/INI1; SNF5L1; Snr1; Sucrose nonfermenting yeast homolog like 1; sucrose nonfermenting, yeast, homolog-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 isoform a; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1a; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1-A; SWI/SNF-related matrix-associated protein; SWI10; SWNTS1; Transcription factor TYE4; Transcription regulatory protein SNF5; TYE4; Unknown (protein for MGC:138029); zgc:92517 | |
Rabbit | |
Protein A | |
RUO | |
20587, 361825, 6598 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.