Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCC1/BAF155, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A301020AM

 View more versions of this product

Catalog No. 50-155-9037


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and contains a predicted leucine zipper motif typical of many transcription factors.
Specifications

Specifications

SMARCC1/BAF155
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
Q92922
BAF155, CRACC1, Rsc8, SRG3, SWI3, BRG1-associated factor 155; SWI/SNF complex 155 kDa subunit; SWI/SNF complex subunit SMARCC1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 1; chromatin remodeling complex BAF155 subunit; mammalian chromatin remodeling complex BRG1-associated factor 155
Rabbit
IgG
100 μl
4° C
Primary
6599
Western Blot
Unconjugated
SMARCC1
liquid
SMARCC1
Between 750 and 800
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit