Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCC1 Mouse anti-Human, Clone: PCRP-SMARCC1-1F1, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen SMARCC1
Clone PCRP-SMARCC1-1F1
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122719
View Documents
Novus Biologicals
NBP30762310UG
100 μg
Each for $352.89
Add to cart
 
NB122720
View Documents
Novus Biologicals
NBP30762320UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

SMARCC1 Monoclonal antibody specifically detects SMARCC1 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

SMARCC1
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
6599
IgG2b
Primary
Store at 4°C.
PCRP-SMARCC1-1F1
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Human
BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3
Recombinant full-length human SMARCC1 protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit