Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCD3 Monoclonal Antibody (OTI7H8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF811108


Description
SMARCD3 Monoclonal antibody specifically detects SMARCD3 in Human samples. It is validated for Western BlotSpecifications
SMARCD3 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8N4V1 | |
Mouse | |
Protein A/G | |
RUO | |
93380 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI7H8 | |
Unconjugated | |
MMGT1 | |
1500001J14Rik; 2210409C08Rik; 60 kDa BRG-1/Brm-associated factor subunit C; BAF60C; BRG1-associated factor 60C; chromatin remodeling complex BAF60C subunit; CRACD3; mammalian chromatin remodeling complex BRG1-associated factor 60C; mBAF60c; MGC111010; Rsc6p; Smarcd3; SWI/SNF complex 60 kDa subunit C; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3, isoform 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 3; Swp73-like protein | |
Full length human recombinant protein of human MMGT1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction