Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ SMART 3™ Colorimeter Reagent Systems
SMART 3™ Colorimeter Reagent Systems for use in water quality testing and environmental studies.
Supplier: LaMotte™ 3653SC
Description
Choose from the following test factors:
- Zinc Laboratory Reagent (LR)
- Manganese
- Aluminum
- Ozone
- Cyanide
- Phosphate
- Dissolved Oxygen
- Chromium Hexavalent
- Ammonia Nitrogen LR
Specifications
2% FS | |
Water testing and environmental studies | |
50 | |
Ascorbic acid reduction |
Phosphate LR | |
SMART colorimeter | |
0 to 3 ppm | |
Phosphate Reagent System |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction