Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ SMART 3™ Colorimeter Reagent Systems

Shop All LaMotte Company Products


Click to view available options
Analytes:
Ammonia Nitrogen
Copper
Cyanide
Dissolved Oxygen
Iron
Manganese LR
Nitrite
Phosphate HR
Phosphate LR
Silica HR
Sulfate
Zinc HR
pH
Range:
0 to 0.5 ppm
0 to 0.7 ppm
0 to 0.8 ppm
0 to 10 ppm
0 to 100 ppm
0 to 3 ppm
0 to 5 ppm
0 to 6 ppm
0 to 70 ppm
0 to 75 ppm
0.05 to 1 ppm
5 to 9.5 ppm
No. of Tests:
20
25
50
100
Description
Choose from the following test factors:
- Zinc Laboratory Reagent (LR)
- Manganese
- Aluminum
- Ozone
- Cyanide
- Phosphate
- Dissolved Oxygen
- Chromium Hexavalent
- Ammonia Nitrogen LR
Specifications
Specifications
| Accuracy | 2% FS |
| Analytes | Sulfate |
| For Use With (Application) | Water testing and environmental studies |
| For Use With (Equipment) | SMART colorimeter |
| No. of Tests | 50 |
| Range | 0 to 100 ppm |
| Test Method | Barium chloride |
| Type | Sulfate Reagent System |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction