Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte SMART Spectro Spectrophometer and SMART 3 Colorimeter Reagent Systems
$22.25 - $140.00
Specifications
For Use With (Application) | Water testing and environmental studies |
---|---|
For Use With (Equipment) | SMART colorimeter |
Catalog Number | Mfr. No. | Product Type | Range | No. of Tests | Test Method | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Range | No. of Tests | Test Method | Price | Quantity & Availability | ||
S19545D
![]() |
LaMotte
3648SC |
Iron Reagent system | 0 to 6ppm | 50 | Bipyridyl |
Each for $53.00
|
|
S19545G
![]() |
LaMotte
3650SC |
Nitrite Reagent System | 0 to 0.8ppm | 20 | Diazotization or coupling |
Each for $59.00
|
|
S19545A
![]() |
LaMotte
365901SC |
Ammonia Nitrogen Test Kit | 0.05 to 1ppm | 25 | Salicylate |
Each for $63.00
|
|
S19545J
![]() |
LaMotte
3665SC |
Sulfate Test Kit | 0 to 100ppm | 50 | Barium chloride |
Each for $38.50
|
|
S19545H
![]() |
LaMotte
3653SC |
Phosphate Test Kit | 0 to 3ppm | 50 | Ascorbic acid reduction |
Each for $49.50
|
|
S19545C
![]() |
LaMotte
3646SC |
Copper Test Kit | 0 to 6ppm | 100 | Diethyldithio-carbmate |
Each for $22.25
|
|
S19545P
![]() |
LaMotte
370001SC |
pH Test Kit | 5 to 9.5ppm | 100 | Colorimetric |
Each for $64.75
|
|
S95109
![]() |
LaMotte
3687SC |
Silica, High Range | 0 to 75ppm | 50 | Silicomolybdate |
Each for $55.00
|
|
S95010
![]() |
LaMotte
3668SC |
Iron | 0 to 5.0ppm | 50 | 1,10 Phenanthroline |
Each for $58.75
|
|
S94995
![]() |
LaMotte
366001SC |
Cyanide | 0 to 0.5ppm | 50 | Pyridine-Barbituric Acid |
Each for $128.00
|
|
S94999
![]() |
LaMotte
3688SC |
Dissolved Oxygen | 0 to 10ppm | 100 | Winkler Colorimetric |
Each for $53.00
|
|
S95104
![]() |
LaMotte
3655SC |
Phosphate, High Range | 0 to 70ppm | 50 | Vanadomolybdophosphoric Acid |
Each for $28.50
|
|
S95012
![]() |
LaMotte
365801SC |
Manganese, Low Range | 0 to 0.7ppm | 50 | PAN |
Each for $79.50
|
|
S95115
![]() |
LaMotte
366701SC |
Zinc, Low Range | 0 to 3ppm | 50 | Zincon |
Each for $140.00
|
|
S95108
![]() |
LaMotte
3664SC |
Silica, Low Range | 0 to 0.4ppm | 100 | Heteropoly blue |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S19545N
![]() |
LaMotte
3649SC/DEL |
Nitrate Reagent System | 0 to 3ppm | 20 | Cadmium reduction |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
Reagents for use in water testing and environmental studies.
Specifications
Water testing and environmental studies | |
SMART colorimeter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title