Learn More
LaMotte™ SMART 3™ Colorimeter Reagent Systems
SCI_ED
SMART 3™ Colorimeter Reagent Systems for use in water quality testing and environmental studies.
Supplier:  LaMotte™ 370001SC
 View more versions of this product
Catalog No. S19545P
Add to Cart
Description
Description
Choose from the following test factors:
  • Zinc Laboratory Reagent (LR)
  • Manganese
  • Aluminum
  • Ozone
  • Cyanide
  • Phosphate
  • Dissolved Oxygen
  • Chromium Hexavalent
  • Ammonia Nitrogen LR
Specifications
Specifications
pH
SMART colorimeter
5 to 9.5 ppm
pH Reagent System
Water testing and environmental studies
100
Colorimetric
Product Suggestions
Product Suggestions