Learn More
LaMotte™ SMART 3™ Colorimeter Reagent Systems
SMART 3™ Colorimeter Reagent Systems for use in water quality testing and environmental studies.
Supplier: LaMotte™ 366001SC
Description
Choose from the following test factors:
- Zinc Laboratory Reagent (LR)
- Manganese
- Aluminum
- Ozone
- Cyanide
- Phosphate
- Dissolved Oxygen
- Chromium Hexavalent
- Ammonia Nitrogen LR
Specifications
Cyanide | |
SMART colorimeter | |
0 to 0.5 ppm | |
Cyanide Reagent System |
Water testing and environmental studies | |
50 | |
Pyridine-Barbituric Acid |