Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Smart-Tracker™ Wireless Data Module Accessories
Maximize Thermo Scientific™ Smart-Tracker™ with additional accessories or personalize your Smart-Tracker with a series of colored bands.
Supplier: Thermo Scientific™ ST501509
Description
Maximize Thermo Scientific™ Smart-Tracker™ with additional accessories or personalize your Smart-Tracker with a series of colored bands.
Specifications
Rings | |
Wireless Monitoring Solution | |
0.24 x 0.63 x 1.02 in. (6 x 16 x 26 mm) [Shipping Dimensions] | |
0.2 kg |
Set of 10 rings (all colors) | |
Smart-Tracker™ Module | |
0.44 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction