Learn More
Medschenker™ Smart Transport Medium & AccuSwab Nasopharyngeal Bundle
For collection and transport of viruses, chlamydia, mycoplasma, and ureaplasma
$3750.00 - $4169.35
Specifications
Cap Lid Type Screw
Material Plastic (STM), Nylon Fiber (Swab)
View More Specs
Products 2
Catalog Number Mfr. No. Fill Volume Price Quantity & Availability  
Catalog Number Mfr. No. Fill Volume Price Quantity & Availability  
23-000-005
View Documents
Medschenker™
STM30AC10B
3 mL
Case of 1000 for $4,169.35
Add to cart
23-000-004
View Documents
Medschenker™
STM15AC10B
1.5 mL
Case of 1000 for $3,750.00
Add to cart
Description
Description
  • For collection, transport, maintenance, and long term freeze storage of viruses, chlamydia, mycoplasma, and ureaplasma
  • Medschenker™ STM includes antibiotics to help inhibit bacteria and fungal flora in the sample for the most accurate results
  • Medschenker™ STM is a stand-up and shatterproof tube enabling quality centrifugation of the sample
  • Patient sample material and particles are rapidly released and dispersed
  • It maintains organisms viability for 48 hours at room or refrigerated temperature
  • MedSchenker™ STM features a new cap-lock system for secure transport without the risk of leaking or spilling
  • Designed for perfect bend without creating additional irritation for the patient
  • Electron beam sterilized and individually wrapped in pouches to ensure 100% sterility
  • Nylon fiber flocked tip that is more comfortable and less intrusive than traditional polyester swabs
  • Developed out of extensive collaboration with physicians and clinical microbiologist
  • This superior design allows for a quick sample uptake and elution of more than 90% of the sample
Specifications
Specifications
Screw
Plastic (STM), Nylon Fiber (Swab)