Thermo Scientific™ Smart-Vue™ Client Server Software
Automate monitoring of vital laboratory equipment parameters with Thermo Scientific™ Smart-Vue™ Client Server Software and enjoy greater peace of mind.
Manufacturer: Thermo Scientific™ SV625500LSB
 View more versions of this product
Catalog No. 13-987-218
  Check Availability
Add to Cart
Disclaimers:
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
    Specifications
    Certifications/Compliance Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
    Description Smart-Vue client server software upgrade version 1.0 to 2.0, up to 99 sensors
    Includes Software drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
    Automate monitoring of vital laboratory equipment parameters with Thermo Scientific™ Smart-Vue™ Client Server Software. Find peace of mind knowing your work is fully monitored 24/7 with immediate notifications reaching you even when you are away from the lab. This software provides a plug and play solution with an intuitive interface.
    Intelligent Plug & Play Simplicity
    • Installation Wizard for simplified, step-by-step process
    • Service Discovery Protocol (SDP) enables every module to send signal to receiver—identifies most efficient communication route based on RF signal strength
    • Module automatically establishes new path to receiver if no communication during 6-hour period
    • Back-up system with memory capacity up to 3000 points, with uninterruptible power for continuous data logging
    Intuitive Dashboard
    • Daily usage is simplified
    • Single-click export of reports to Microsoft™ Excel™, Word™ or Adobe™ PDF formats
    • Flexible creation of graphs summarizing data from individual or multiple sensors
    • Visual depiction of RF signal strength an QoS as monitored by each module
    Data Traceability & Security
    • Secure, customized levels of user access
    • Audit trails
    • Archived alarms
    • Password protection
    • Secure digital transmission and data back-ups
    Numerous Configuration Capabilities
    • Combines RF and TCP/IP network technologies
    • Share equipment data over entire network instantly
    • Download simple plug-in to visualize equipment data readings from any workstation
    Compatible with:
    Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
    Recommended for:
    Ideal for use in highly controlled environments, including pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories.
    Ordering Information:
    Availability of Smart-Vue products vary by country. Please ask your local sales representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
  • Description & Specifications
Related Products
Product Content Feedback