Learn More
Research Products International CorpSupplier Diversity Partner Disposable smartSpatula, 210mm Long, Blue, 300 per Package DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 249902
This disposable SmartSpatula provides the most innovative design for a new multi-purpose lab tool. Features a hollow core handle with both a scoop and spatula end for convenient transfer of collection of materials. Can also be used to stir solutions. Flexible ends become shatter resistant after freezing in liquid nitrogen and readly form to an object to scrape or collect samples. Manufactured from chemical resistant polypropylene and can be autoclaved.
Catalog No. 50-476-569