Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ SmartWall Upright

Create a unique storage and workstation system for underutilized wall space with SmartWall Uprights.

$83.40 - $285.00

Specifications

Autoclavable Not Autoclavable
Depth (English) 1.875 in.
Color Gray
Material Epoxy-coated Steel
For Use With (Application) All Laboratory and Clean Room Non-Autoclavable Applications
View More Specs

Products 5
Catalog Number Mfr. No. Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Price Quantity & Availability  
13-460-313
View Documents
Metro™
SWU15K4
15 in.
Each for $83.40
Add to cart
 
13-460-312
View Documents
Metro™
SWU30K4
30 in.
Each for $97.70
Add to cart
 
13-460-311
View Documents
Metro™
SWU45K4
45 in.
Each for $113.70
Add to cart
 
13-460-310
View Documents
Metro™
SWU72K4
72 in.
Each for $272.00
Add to cart
 
13-460-314
View Documents
Metro™
SWU85K4
85 in.
Each for $285.00
Add to cart
 
Description

Description

  • Built-in slots allow shelf supports and grids to attach without tools or hardware.
  • 1.5 in. (38 mm) slot spacing allows shelves and grids to be adjusted on 1.5 in. (38 mm) increments.
  • Designed for medium-duty applications.
  • Two uprights are required for a stand-alone unit.
  • Each part number is one individual upright.
Specifications

Specifications

Not Autoclavable
Gray
All Laboratory and Clean Room Non-Autoclavable Applications
1.125 in.
1.875 in.
Epoxy-coated Steel
SmartWall Shelving
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit