Learn More
Aviva Systems Biology Corp SMC3 Antibody - C-terminal region (ARP47590_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47590_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMC3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SMC3. Alternate names: BAM, BMH, CDLS3, CSPG6, HCAP, SMC3L1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59600