Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen SMC4
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml, Flow Cytometry
Applications Western Blot, Flow Cytometry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056394T
SDP
View Documents
Novus Biologicals
NB10056394SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056394
SDP
View Documents
Novus Biologicals
NB10056394
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

SMC4 Polyclonal specifically detects SMC4 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

SMC4
Western Blot 1-3 ug/ml, Flow Cytometry
Polyclonal
Purified
RUO
Human
NP_001099000
10051
A synthetic peptide corresponding to amino acids 281-297 of human CAP-C. Genbank accession no. NP_001002800.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Flow Cytometry
Unconjugated
Rabbit
Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
PBS containing 0.2% gelatin with 0.05% Sodium Azide
CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog
SMC4
IgG
Protein G purified
Cross reacts with Human. Predicted to react with Mouse, Rat, Xenopus and Chicken.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.