Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA540133
Description
Recommended positive controls: 293T, A431, HeLa, and HepG2 cells. Predicted reactivity: Mouse (92%), Rat (92%), Rhesus Monkey (96%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SMC4 Polyclonal antibody specifically detects SMC4 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
SMC4 | |
Polyclonal | |
Unconjugated | |
Smc4 | |
2500002A22Rik; C79747; Capc; CAP-C; chromosome-associated polypeptide C; hCAP-C; SMC protein 4; SMC4; SMC-4; SMC4 structural maintenance of chromosomes 4-like 1; SMC4L1; structural maintenance of chromosomes 4; structural maintenance of chromosomes protein 4; XCAP-C homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10051 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9NTJ3 | |
Smc4 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SMC4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction