Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMCP Monoclonal Antibody (5C10D8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531914
Description
MA5-31914 has been tested in indirect ELISA.
Involved in sperm motility. Its absence is associated with genetic background dependent male infertility. Infertility may be due to reduced sperm motility in the female reproductive tract and inability to penetrate the oocyte zona pellucida.
Specifications
SMCP | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P49901 | |
SMCP | |
Purified recombinant fragment of human SMCP (AA: FULL(1-116)) expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Western Blot, Immunocytochemistry | |
5C10D8 | |
Unconjugated | |
SMCP | |
HSMCSGEN1; MCS; MCSP; mitochondrial capsule selenoprotein; SMCP; sperm mitochondria associated cysteine rich protein; sperm mitochondria-associated cysteine-rich protein; sperm mitochondrial-associated cysteine-rich protein; testicular tissue protein Li 119; testicular tissue protein Li 161 | |
Mouse | |
Protein G | |
RUO | |
4184 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction