Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMCP Mouse anti-Human, DyLight 755, Clone: 5C10A2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237278IR

Catalog No. NBP237278IR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

SMCP
5C10A2
Western Blot, ELISA
Purified
SMCP
Purified recombinant fragment of human SMCP (AA: 1-116) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
HSMCSGEN1, MCSMGC26305, MCSPMGC26519, mitochondrial capsule selenoprotein, sperm mitochondria-associated cysteine-rich protein, sperm mitochondrial-associated cysteine-rich protein
Mouse
IgG2b
0.1 ml
Primary
4184
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit