Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMCP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548460

Catalog No. PA548460

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in sperm motility. Its absence is associated with genetic background dependent male infertility. Infertility may be due to reduced sperm motility in the female reproductive tract and inability to penetrate the oocyte zona pellucida.
Specifications

Specifications

SMCP
Polyclonal
PBS with 0.09% sodium azide
P49901
Sperm mitochondrial-associated cysteine-rich protein, SMCP, MCS, MCSP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4184
Western Blot
Unconjugated
SMCP
Liquid
SMCP
KLH conjugated synthetic peptide between 68-97 amino acids from the C-terminal region of human SMCP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit