Learn More
SMCR7/MID49 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 164131AP
Catalog No. 50-173-0928
Add to cart
Description
Description
This antibody recognizes isoform1 49 kDa only.
This gene encodes an outer mitochondrial membrane protein that functions in the regulation of mitochondrial morphology. It can directly recruit the fission mediator dynamin-related protein 1 (Drp1) to the mitochondrial surface. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
SMCR7/MID49
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5NCS9, Q96C03
MID49, MIEF2, SMCR7
Rabbit
IgG
150 μL
-20°C
Primary
125170, 237781
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MIEF2
Liquid
Mief2
SMCR7/MID49 Fusion Protein Ag9531
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse