Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMCR7L Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen SMCR7L
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095587
View Documents
Novus Biologicals
NBP303717100UL
100 μg Each for $320.71
Add to cart
 
NB095588
View Documents
Novus Biologicals
NBP30371720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

SMCR7L Polyclonal antibody specifically detects SMCR7L in Human samples. It is validated for Western Blot
Specifications

Specifications

SMCR7L
Unconjugated
Rabbit
Human
dJ1104E15.3, FLJ20232, HSU79252, MID51, Smith-Magenis syndrome chromosomal region candidate gene 7 protein-like, Smith-Magenis syndrome chromosome region, candidate 7-like
Recombinant fusion protein containing a sequence corresponding to amino acids 40-280 of human SMCR7L (NP_061881.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
54471
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit