Smith Medical Saf-T-Lance™ Safety Lancets

Retractable safety technology for capillary blood sampling

Manufacturer: Smiths Medical PM, Inc. 1014

 View more versions of this product

Catalog No. 22-024-982

  Check Availability
Add to Cart

    Specifications

    Color Green
    Flow Type Normal
    For Use With (Application) For capillary blood sampling
    Quantity 3600 Case
    Shelf Life 5 Year(s)
    Product Type Safety Lancet
    Needle Gauge 21g

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit