Smith Medical Saf-T-Lance™ Safety Lancets

Retractable safety technology for capillary blood sampling

Automatic retraction reduces needlestick injuries and cross-contamination exposure while providing a quick, gentle puncture. Needle is shielded before and after use.

$1128.90 - $1356.14

Specifications

For Use With Application For capillary blood sampling
Product Type Safety Lancet
Quantity 3600 Case
Shelf Life 5 Year(s)
View More Specs
  • Products 5
  • Description & Specifications
Products
Catalog Number Mfr. No. Color Price Quantity & Availability    

22-024-971

Smith Medical
1020
Blue Case of 3600 for $1,356.14
Check Availability
Add to Cart

22-024-981

SMITH MEDICAL
1008
Yellow Case of 3600 for $1,356.14
Check Availability
Add to Cart

22-025-003

SMITH MEDICAL
1005
Purple Case of 3600 for $1,128.90
Check Availability
Add to Cart

22-024-982

SMITH MEDICAL
1014
Green Case of 3600 for $1,356.14
Check Availability
Add to Cart

22-025-005

SMITH MEDICAL
1028
N/A N/A N/A  
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Descriptions & Specifications

Related Products